Anti-IgG F(ab')2 Goat Polyclonal Antibody (Biotin)

Proveedor: Rockland Immunochemicals
709-1618
ROCK709-1618EA 490 EUR
ROCK709-1618
Anti-IgG F(ab')2 Goat Polyclonal Antibody (Biotin)
Anticuerpos
This product is designed for immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms.

Recommended Dilutions: ELISA: 1:500,000; Western Blot: 1:5000 - 1:40000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: F(ab')2
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR