Anti-IgG (H+L) Goat Polyclonal Antibody (Peroxidase)

Proveedor: Rockland Immunochemicals
710-103-019
ROCK710-103-019EA 436 EUR
ROCK710-103-019
Anti-IgG (H+L) Goat Polyclonal Antibody (Peroxidase)
Anticuerpos
Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 500 µl
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR