Anti-IgG F(ab')2 Goat Polyclonal Antibody (Texas Red®)

Proveedor: Rockland Immunochemicals
710-1931
ROCK710-1931EA 562 EUR
ROCK710-1931
Anti-IgG F(ab')2 Goat Polyclonal Antibody (Texas Red®)
Anticuerpos
Secondary Goat Anti-F(ab')2 Reacts with Mouse

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: F(ab')2
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

ESPECIFICACIONES

Simbolo antígeno IgG F(ab')2
Environmentally Preferable
Tipo de anticuerpo Secundario
Clonalidad Polyclonal
Conjugado Texas Red®
Reactividad Mouse
Huésped Goat
Isotipo
Western blot Yes
ELISA Yes
InmunoQuímica Yes
Precipitación inmunológica Yes
FluorescenciaInmunológica Yes
Cambio de gel Yes
Citometría de flujo Yes
ChIP Yes
Neutralización Yes
Fragmento de anticuerpo F(ab')2
Inmunógeno Mouse IgG F(ab')2 fragment
Formato Antibody fraction
Forma Lyophilized
Purification purified by immunoaffinity chromatography
Concentración 1,0 mg/ml by UV absorbance at 280 nm
Storage buffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR