Chemiluminescent super sensitive HRP substrate, FemtoMax™
Proveedor: Rockland Immunochemicals
FEMTOMAX-020 FEMTOMAX-110
ROCKFEMTOMAX-020EA 260 EUR
ROCKFEMTOMAX-020 ROCKFEMTOMAX-110
Chemiluminescent super sensitive HRP substrate, FemtoMax™
Reactivos para electroforesis Western Blotting Reagents
FemtoMax™ is an extremely sensitive, non radioactive, and enhanced luminol-based chemiluminescent substrate for the detection of horseradish peroxidase (HRP). It allows for the detection of femtogram (10 to 15) amounts of antigen using photographic film or other imaging methods including highly sensitive CCD cameras.
  • Clear, colourless, odourless liquid
  • Blots can be repeatedly exposed to X-ray film
  • Optimal results or stripped of detection reagents and re-probed
FemtoMax™ finds use in both Western blotting and enzyme immunoassay (EIA).
Información de suministro: Supplied as two component system- luminol reagent (reagent A) and reaction buffer (reagent B).
Precaución: Store at 4 °C prior to opening. Protect from moisture and light.