Latex gloves, Gentle Skin® grip
Proveedor: MEDITRADE
1221G-S
ROES1221G-SEA 17.7 EUR
ROES1221G-S
Latex gloves, Gentle Skin® grip
Guantes
These disposable latex gloves are ideal for dental surgery and any other environments where the safe, non-slip handling of instruments and machinery is a top priority.
  • Optimal, secure grip
  • Hypoallergenic: powder-free, very low protein/accelerator content
  • Very easy to pull on
  • Micro-roughened for responsive, outstanding touch sensitivity
  • Highly elastic and tear-proof
Certificaciones: EN 455, EN 374; Cat. III; AQL 1,5
Food contact approved