XTRA™ Carts
Proveedor: Rubbermaid Commercial Products
FG409400GRAY FG409100BLA FG409300BLA FG409600BLA FG409100OWHT
RUBBFG409400GRAYEA 1210 EUR
RUBBFG409400GRAY RUBBFG409100BLA 129-1833 129-1832 RUBBFG409100OWHT
XTRA™ Carts
Carritos
Durable and attractive storage and transportation carts for front and back-of house applications.
  • Metal uprights and moulded shelves engineered for durability and appearance
  • Non-marking 10,2 cm swivel casters
  • Wide shelves hold a variety of equipment and supplies, up to 135 kg
  • Available in various versions
Información accesorios: Accessories are available for a complete system solution for housekeeping in the hospitality industry. Utility bin features a smooth easy-to-clean surface.
Información para pedidos: Attachable plastic container must beMore Product Information