Solvent Blue 38, Practical grade, Sigma-Aldrich®

Proveedor: SIGMA ALDRICH MICROSCOPY

Sinónimos: Direct Blue 86, C.I. 74180, Luxol® Fast Blue MBSN

M-Clarity™ quality level = MQ200

S3382-100G S3382-25G
S3382-100GEA 283 EUR
S3382-100G S3382-25G
Solvent Blue 38, Practical grade, Sigma-Aldrich®
Solvent Blue 38

Solvent Blue 38 is a phthalocyanine stain, used for staining myelin, neurons and glial fibres.


  • Solvent Blue 38 has been used to stain tissue sections to assess remyelination


Solvent Blue 38 exhibits a strong affinity towards bases in the lipoproteins of myelin sheath due to which it is stained blue. Luxol is a registered trademark of Rohm and Haas Co.

Formula: C32H12CuN8Na2O6S2
Temperatura de almacenaje: Room Temperature
Número MDL: MFCD00071424
Núm. CAS: 1328-51-4

Order Now

Resultados test especificaciones

Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Nitrogen 14.5 - 16.5 %
Copper (Cu) 3.5 - 5.5 %
Solubility (Colour)
1 mg/ml, methanol
Dark Blue


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR