Cohesive sampler
Proveedor: Sampling Systems
2000S-01
SAMP2000S-01EA 2390 EUR
SAMP2000S-01
Cohesive sampler
Muestreadores Muestreadores de sólidos
This cohesive sampler is designed for powders.
  • For taking single point samples
  • Handle and four sample cells
  • 0,25, 0,50, 0,75, 1,0 ml unit doses
Used throughout the pharmaceutical industry from sticky (or cohesive) powders.
Información para pedidos: A Nylon bristle can be ordered separately.
Información de suministro: In a case.