Weighing PharmaScoop
Proveedor: Sampling Systems
A376-500
SAMPA376-500EA 221 EUR
SAMPA376-500
Weighing PharmaScoop
Palas
Designed for convenient weighing of powders.
  • Made from high-quality 316L stainless steel
  • Flat bottom design enables quick and easy weighing
  • Widely used across both food and pharmaceutical industries
  • Available from stock
  • Fast delivery
The weighing PharmaScoop is ideal for quickly weighing out powders due to its' flat bottom design. It is made from high-quality 316L stainless steel and features a crevice-free design, making it extremely easy to clean.