Cubis® II MCA 225S Ultra-High Resolution Semi-Micro Balances
Proveedor: Sartorius Balances
MCA225S3S014 MCA225S3CE14H MCA225S3S0124I MCA225S3CE3M MCA225S3CE12H MCA225S3CE123M MCA225S3S0I MCA225S3S014H MCA225S3S012I MCA225S3CEM MCA225S3CE4 MCA225S3CE2H MCA225S3CE13H MCA225S3CE34I MCA225S3S04I MCA225S3CE234I MCA225S3CE23M MCA225S3CE34M MCA225S3S0124M MCA225S3S034H MCA225S3S01M MCA225S3CE14M MCA225S3CE24 MCA225S3CE2I MCA225S3CE124 MCA225S3S013M MCA225S3S03I MCA225S3CE14I MCA225S3S04H MCA225S3S023I MCA225S3S099M MCA225S3CE99I MCA225S3S023H MCA225S3S014M MCA225S3CE34H MCA225S3CE3I MCA225S3S012 MCA225S3CE99M MCA225S3CE23H MCA225S3S0123M MCA225S3CE13M MCA225S3CE12I MCA225S3CE4M MCA225S3S0123H MCA225S3CE234M MCA225S3S0123 MCA225S3S013 MCA225S3CE13I MCA225S3S024H MCA225S3S034M MCA225S3CE234H MCA225S3S023M MCA225S3S0124H MCA225S3S014I MCA225S3S012H MCA225S3CE23 MCA225S3CE14 MCA225S3CE2 MCA225S3CE4I MCA225S3S03M MCA225S3CE3H MCA225S3CE23I MCA225S3S023 MCA225S3S0124 MCA225S3CE34 MCA225S3CE123H MCA225S3CE12M MCA225S3S0234M MCA225S3CE123 MCA225S3CEI MCA225S3S024M MCA225S3S03 MCA225S3S0234I MCA225S3S04 MCA225S3S024 MCA225S3S0123I MCA225S3S024I MCA225S3CE123I MCA225S3S0234H MCA225S3S02I MCA225S3CE234 MCA225S3CE3 MCA225S3CE124H MCA225S3S012M MCA225S3CE124M MCA225S3CE2M MCA225S3S01I MCA225S3CE4H MCA225S3CE12 MCA225S3S04M MCA225S3CE1I MCA225S3S02 MCA225S3S034I MCA225S3S034 MCA225S3CE1M MCA225S3S013H MCA225S3S0M MCA225S3S02M MCA225S3CE13 MCA225S3CE24H MCA225S3CE24M MCA225S3S0234 MCA225S3S099I MCA225S3S02H MCA225S3CE24I MCA225S3CE124I MCA225S3S03H MCA225S3S013I
SARBMCA225S3S014EA 23355 EUR
SARBMCA225S3S014 SARBMCA225S3CE14H SARBMCA225S3S0124I SARBMCA225S3CE3M SARBMCA225S3CE12H SARBMCA225S3CE123M SARBMCA225S3S0I SARBMCA225S3S014H SARBMCA225S3S012I SARBMCA225S3CEM SARBMCA225S3CE4 SARBMCA225S3CE2H SARBMCA225S3CE13H SARBMCA225S3CE34I SARBMCA225S3S04I SARBMCA225S3CE234I SARBMCA225S3CE23M SARBMCA225S3CE34M SARBMCA225S3S0124M SARBMCA225S3S034H SARBMCA225S3S01M SARBMCA225S3CE14M SARBMCA225S3CE24 SARBMCA225S3CE2I SARBMCA225S3CE124 SARBMCA225S3S013M SARBMCA225S3S03I SARBMCA225S3CE14I SARBMCA225S3S04H SARBMCA225S3S023I SARBMCA225S3S099M SARBMCA225S3CE99I SARBMCA225S3S023H SARBMCA225S3S014M SARBMCA225S3CE34H SARBMCA225S3CE3I SARBMCA225S3S012 SARBMCA225S3CE99M SARBMCA225S3CE23H SARBMCA225S3S0123M SARBMCA225S3CE13M SARBMCA225S3CE12I SARBMCA225S3CE4M SARBMCA225S3S0123H SARBMCA225S3CE234M SARBMCA225S3S0123 SARBMCA225S3S013 SARBMCA225S3CE13I SARBMCA225S3S024H SARBMCA225S3S034M SARBMCA225S3CE234H SARBMCA225S3S023M SARBMCA225S3S0124H SARBMCA225S3S014I SARBMCA225S3S012H SARBMCA225S3CE23 SARBMCA225S3CE14 SARBMCA225S3CE2 SARBMCA225S3CE4I SARBMCA225S3S03M SARBMCA225S3CE3H SARBMCA225S3CE23I SARBMCA225S3S023 SARBMCA225S3S0124 SARBMCA225S3CE34 SARBMCA225S3CE123H SARBMCA225S3CE12M SARBMCA225S3S0234M SARBMCA225S3CE123 SARBMCA225S3CEI SARBMCA225S3S024M SARBMCA225S3S03 SARBMCA225S3S0234I SARBMCA225S3S04 SARBMCA225S3S024 SARBMCA225S3S0123I SARBMCA225S3S024I SARBMCA225S3CE123I SARBMCA225S3S0234H SARBMCA225S3S02I SARBMCA225S3CE234 SARBMCA225S3CE3 SARBMCA225S3CE124H SARBMCA225S3S012M SARBMCA225S3CE124M SARBMCA225S3CE2M SARBMCA225S3S01I SARBMCA225S3CE4H SARBMCA225S3CE12 SARBMCA225S3S04M SARBMCA225S3CE1I SARBMCA225S3S02 SARBMCA225S3S034I SARBMCA225S3S034 SARBMCA225S3CE1M SARBMCA225S3S013H SARBMCA225S3S0M SARBMCA225S3S02M SARBMCA225S3CE13 SARBMCA225S3CE24H SARBMCA225S3CE24M SARBMCA225S3S0234 SARBMCA225S3S099I SARBMCA225S3S02H SARBMCA225S3CE24I SARBMCA225S3CE124I SARBMCA225S3S03H SARBMCA225S3S013I
Cubis® II MCA 225S Ultra-High Resolution Semi-Micro Balances
Balanzas
The Cubis® II laboratory balances are modular, therefore they allow to choose between applications and configurations which suit the best to the needs. These balances can be configured at the level of display, draftshields, software applications and hardware functions.
  • Draft shield (D) for high capacity micro balances with upgradable hardware
  • Superior weighing performance
  • No more issues with charged samples
  • Cleaning process guidance
  • Hardware upgradeability, compliance and data integrity
  • Easy to use with motorised inner draft shield
MCA display - 7"More Product Information