Micro laboratory balances, Cubis® II MCA
Proveedor: Sartorius Balances
MCA10.6S2SF99 MCA10.6S2SM99 MCA10.6S2S00-F MCA6.6S2S00-F MCA3.6P2S00-F MCA10.6S2S00-M MCA6.6S2S00-M MCA3.6P2S00-M MCA6.6S2S00F99 MCA3.6P2S00F99 MCA6.6S2S00M99 MCA3.6P2S00M99 MCA66S2S00MQP1
SARBMCA10.6S2SF99EA 42390 EUR
SARBMCA10.6S2SF99 SARBMCA10.6S2SM99 SARBMCA10.6S2S00-F SARBMCA6.6S2S00-F SARBMCA3.6P2S00-F SARBMCA10.6S2S00-M SARBMCA6.6S2S00-M SARBMCA3.6P2S00-M SARBMCA6.6S2S00F99 SARBMCA3.6P2S00F99 SARBMCA6.6S2S00M99 SARBMCA3.6P2S00M99 611-5263
Micro laboratory balances, Cubis® II MCA
Balanzas
Cubis® II MCA micro balances with advanced user interface offer readability of 1 μg and a maximum capacity between 3,1 and 10,1 g. Used to determine the exact weight of very small sample quantities.
  • Modular premium balances with thousands of hardware and software configurations and adaptable software
  • Seamless connectivity
  • Long-term flexibility
  • Submission-ready data
  • Governance through user management
  • Integration of SOPs
  • Intuitive applications
  • Status center notifications
Weighing in a regulated environment can be complex, with stringent complianceMore Product Information