Sample tube with ventilation cap
Proveedor: SARSTEDT
55.526.006 62.515.006 62.476.028 62.515.028 55.476.013 62.526.028
SARS55.526.006EA 0 EUR
SARS55.526.006 SARS62.515.006 SARS62.476.028 SARS62.515.028 SARS55.476.013 SARS62.526.028
Sample tube with ventilation cap
Tubos Tubos de ensayos
PS, clear or PP, translucent.
  • PP: good temperature resistance, good chemical resistance
  • PS: highly transparent, well suited to visual measurements
Universal tubes for a wide range of applications.