Blotting membrane, nitrocellulose
Proveedor: Sartorius
11327-41BL
SART11327-41BLEA 412 EUR
SART11327-41BL
Blotting membrane, nitrocellulose
Membranas de transferencia
These nitrocellulose blotting membranes are ideal for Western blotting, DNA blotting, as well as dot or slot blots. They are optimally adapted to all protein blotting systems, such as electro transfer, semi-dry or simple capillary blotting. A high protein binding capacity is combined with low background to give good blocking possibilities. High membrane stability ensures easy handling of the blotting membrane, even when wet.
  • Optimally suited for dot or slot blotting
  • Fast and complete protein transfers
  • Available with 0,22 or 0,45 µm pore size
  • High surface