Vacuum filter holders, Combisart®
Proveedor: Sartorius
16219-CS 16201-CS
SART16219-CSEA 1090 EUR
SART16219-CS SART16201-CS
Vacuum filter holders, Combisart®
Portafiltros
Individual stainless steel filter holders, stand-alone, to be assembled on vacuum flasks or on manifolds, depending on the particular version.
Stainless steel holders differ in their funnel capacity (choice of 100 ml or 500 ml) and in their valve type (part numbers ending with CS feature sterile venting). They are specifically designed for applications in which particles or microorganisms need to be retained on the surface of membrane filters. Their stainless steel frit filter support ensures uniform distribution of the retained particles over the entire surface of the membrane filter.More Product Information