Syringe filter holder, stainless steel
Proveedor: Sartorius
16214
SART16214EA 313 EUR
SART16214
Syringe filter holder, stainless steel
Portafiltros
This syringe filter holder is made from stainless steel. PTFE coated surface on the top provides leak proof sealing, and the chemical compatibility depends upon the filter inserted.
  • Re-usable
  • Resistant to heat