Qualitative filter papers, technical, smooth, grade 3 m/N
Proveedor: Sartorius
FT2-305-580580 FT-4-305-185 FT-4-305-125 FT-3-305-270 FT-3-305-185 FT-3-305-150 FT-4-305-240 FT-3-305-125 FT-4-305-150 FT-3-305-110 FT-3-305-240 FT-4-305-385
516-7100EA 110 EUR
516-7100 SARTFT-4-305-185 SARTFT-4-305-125 SARTFT-3-305-270 SARTFT-3-305-185 515-1592 SARTFT-4-305-240 515-1591 SARTFT-4-305-150 SARTFT-3-305-110 SARTFT-3-305-240 SARTFT-4-305-385
Qualitative filter papers, technical, smooth, grade 3 m/N
Filtros Papeles de filtros Papel de filtro cualitativo
These smooth filter papers are made of refined pulp and linters with more than 95% alpha-cellulose content. The ash content of these filter papers is between 0,1 and 0,15%. They are used for routine analyses like clarification, determination of substances, but also as discs with a centre hole for technical applications. Grades with a wet burst resistance >30 kPa are referred to as wet-strengthened and are therefore suitable for pressure or vacuum filtration.
Medium fast rate of filtration, thin, smooth, increased wet strength. Used for acids; general vacuum or pressure filtrationMore Product Information