ThermoSafe® PUR Insulated Shippers
Proveedor: Sonoco Thermosafe
E89 E90
SCATE89EA 231 EUR
SCATE89 SCATE90
ThermoSafe® PUR Insulated Shippers
Remitentes de envíos y contenedores
High perfomance polyurethane (PUR) insulated shipper with foam plug closure.
  • PUR offers greater insulating capability when compared to same size EPS container
  • Thick walls and outer corrugate provides protective shock absorption and cushioning during shipping and handling
  • Protects highly temperature sensitive shipments
  • Economical and lightweight
Rigid, one-piece, three-layer laminate construction provides excellent durability. Proprietary process laminates insulation core to inner and outer corrugate walls for superior strength and temperature controlMore Product Information