Serum vials, borosilicate glass
Proveedor: DWK Life Sciences
SP62113D-312 6212D-20 SP62113D-2 SP62121D-10 SP62121D-5 SP62121U-10 4042467
SCERSP62113D-312EA 44.4 EUR
SCERSP62113D-312 SCER6212D-20 SCERSP62113D-2 SCERSP62121D-10 SCERSP62121D-5 SCERSP62121U-10 WHEA4042467
Serum vials, borosilicate glass
Viales
These serum vials are ideally suited for the handling, containment, and storage of a variety of liquids, including reagents, vaccines, blood plasma, culture media, chromatography samples.
  • Special design provides extra strength for freeze-drying applications
  • Lighter in weight, with more closely held tolerances and uniformity of glass
  • Special blowback design provides ease of use for lyophillization stoppers
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.