Microfiltration assembly, with solvent pick-up adapter, ULTRA-WARE®, KIMBLE®
Proveedor: DWK Life Sciences
SP953820-1047
SCERSP953820-1047EA 1320 EUR
SCERSP953820-1047
Microfiltration assembly, with solvent pick-up adapter, ULTRA-WARE®, KIMBLE®
Filtros Embudos de filtros de membrana
This microfiltration system manufactured from 33 expansion, low extractable borosilicate glass eliminates pouring HPLC solvents into funnels. The solvent pickup adapter draws solvent directly from a reagent bottle through a 47 mm filter and into a filter flask.
  • Support base with 40 to 60 micron porosity glass frit
  • Integral vacuum connection above the drip tip to prevent contamination of the vacuum line with filtrate droplets
After filtration is complete, the HI-VAC valve on the solvent pickup adapter can be turned off for vacuum degassing.
Order Now
ESPECIFICACIONES
Environmentally Preferable
Capacidad 1000 ml