Ultrospec 7500 UV-Visible Spectrophotometer
Proveedor: BIOCHROM
80-2140-60
SCLI80-2140-60EA 12740 EUR
SCLI80-2140-60
Ultrospec 7500 UV-Visible Spectrophotometer
Espectrofotómetros y Fotómetros Espectrofotómetros UV/Vis
A simple to use instrument with advanced performance and functionality, the Ultrospec 7500 is perfect for any busy lab, from academia to GLP manufacturing facilities.
  • Easy to use icon driven touch screen for true 'plug and play' setup
  • Automated 8-cell sample carousel supplied as standard
  • Robust, compact and reliable for general or heavily regulated laboratory use
  • Can be upgraded with Resolution CFR PC control software to comply with 21 CFR Part 11
  • Provides stored methods for DNA, RNA, oligo and protein quantification, as well as checking sample purity