Precision thermometer, stem form ASTM-like 121 C, capillary white enamelled round, blue liquid -ACCU-SAFE, range: +98,6+101,4°C in 0.05°C auxiliary scale: 0°C, total immersion length, total length 320 mm, including DAkkScalibration certificate with 3 1 * 1 UN
Proveedor: Ludwig Schneider GmbH & CO. KG