Precision saccharometers with thermometer and certification
Proveedor: Ludwig Schneider GmbH & CO. KG
3292061S 3292062S 3292063S 3292064S 3292065S 3292066S 3292067S
SCNL3292061SEA 152 EUR
SCNL3292061S SCNL3292062S SCNL3292063S SCNL3292064S SCNL3292065S SCNL3292066S SCNL3292067S
Precision saccharometers with thermometer and certification
Hidrómetros
Final attenuation saccharometers for the brewing industry.
  • Blue liquid filling
  • Reference temperature: 20 °C
Certificaciones: DIN 12790, ISO 387.
Envase: In a square plastic sleeve.