Metal combination electrodes, ScienceLine
Proveedor: SI Analytics
285100075 285102208 285102343 285102351 285102413 285102019 285102002 285102232 285102281 285102090 285102070
662-6122EA 294 EUR
662-6122 SCOI285102208 662-6121 SCOI285102351 SCOI285102413 SCOI285102019 SCOI285102002 SCOI285102232 SCOI285102281 SCOI285102090 662-1371
Metal combination electrodes, ScienceLine
Electrodos Otros electrodos
These combination electrodes with plug head and connection cable are the ideal choice for titrations.