Graduated Pipettes, Type 2, Class AS
Proveedor: DWK Life Sciences
233480608 233481107 233481604 233482309 233482909 233483202 233483408 233483605
SCOT233480608EA 99.4 EUR
SCOT233480608 SCOT233481107 SCOT233481604 SCOT233482309 SCOT233482909 SCOT233483202 SCOT233483408 SCOT233483605
Graduated Pipettes, Type 2, Class AS
Pipetas Pipetas graduadas
Made from soda-lime glass, delivered with batch certificate.
  • Calibrated to deliver (TD, Ex)
  • Type 2: Drain-out, nominal volume at the top (numbering from bottom to the top)
  • Graduation marks and inscriptions in highly durable blue print
  • With cotton plug ending (cotton plugs included)
Certificaciones: Comply with ISO 835.