Safety Waste Collectors
Proveedor: S.C.A.T.
306493 306498 306494 306491 306497 306489 306492 306491-EL 306492-EL 306493-EL 306494-EL 306497-EL 306498-EL 306489-EL
SCTV306493EA 75.6 EUR
SCTV306493 SCTV306498 SCTV306494 SCTV306491 SCTV306497 SCTV306489 SCTV306492 SCTV306491-EL SCTV306492-EL SCTV306493-EL SCTV306494-EL SCTV306497-EL SCTV306498-EL SCTV306489-EL
Safety Waste Collectors
Tapas Tapones de seguridad y para residuos
The Safety Waste Collector connects capillaries and tubes of the HPLC system with the disposal system - suitable for the SymLine benchtop feedthroughs and SymLine FLEX.
  • For collecting capillaries and tubes in a cap on the disposal container.
  • Various number of connections on one cap with standardised GL45 threads
  • Available in electrically conductive (black core) and non-electrically conductive (white core) version
SymLine is the modular disposal system for liquid waste (e.g. solvents, acids, alkalis, etc.). It connects several laboratory workstations,More Product Information