HPLC starter kits, Supply V2.0
Proveedor: S.C.A.T.
399200 399201 399202
SCTV399200EA 288 EUR
SCTV399200 SCTV399201 SCTV399202
HPLC starter kits, Supply V2.0
Tapas Tapones de seguridad y para residuos
Starter kits consisting of SCAT SafetyCaps, blind plugs, fittings and air valves for the supply of HPLC systems.
  • Price advantage compared to ordering piece by piece
  • Suitable for all current HPLC systems
  • Unused connectors can be closed by the blind screws delivered with the set, which make the system universally applicable, even when less connectors are needed
Initial removal equipment that can be used with all common HPLC systems, for the safe extraction of solvents.
Información para pedidos: All set components can be reordered separately.