Tilting mixer shaker, Movil-Tub
Proveedor: SELECTA
7000913
SELE7000913EA 431.66 EUR
SELE7000913
Tilting mixer shaker, Movil-Tub
Agitadores de laboratorio Agitadores Basculantes y de Rotación
Tilting shaker mixer designed with removable anti slide tray made of silicone. Ideal for haematology, chemistry and microbiology applications.
  • Fixed speed