SGE OptChem™ Tapered FocusLiner Inlet Liners
Proveedor: Trajan Scientific and Medical
0926200301 09262003 0926211101 09262111
SGEA0926200301EA 66.1 EUR
SGEA0926200301 SGEA09262003 SGEA0926211101 SGEA09262111
SGE OptChem™ Tapered FocusLiner Inlet Liners
Suministros de cromatografía Inlet Liners de cromatografía
The SGE inlet liner range aims to make it simple for all gas chromatographers to select the correct liner. Choosing the right inlet liner and injection parameters can increase peak areas and reduce detection limits by up to 300%.
  • Easy to choose - colour coded by geometry to simplify your selection
  • Easy to use - contains O-rings so ready to use
  • Bottom taper focuses sample onto the head of the column and minimizes contact with metal parts of the inlet
  • A taper at the top aids in minimizing sample flashback
  • Ensures quartz wool remains in the correct position