Human recombinant Eotaxin (from E. coli)
Proveedor: Shenandoah Biotechnology
100-178-100UG 100-178-1MG 100-178-20UG 100-178-500UG 100-178-5UG
SHBT100-178-100UGEA 453 EUR
SHBT100-178-100UG SHBT100-178-1MG SHBT100-178-20UG SHBT100-178-500UG SHBT100-178-5UG
Human recombinant Eotaxin (from E. coli)
Proteínas y Péptidos
Eotaxin, also known as Eotaxin-1 and CCL11, is a chemokine that recruits eosinophils to sites of allergic inflammation. Eotaxin signals through the G protein-coupled chemokine receptors CCR2, CCR3, and CCR5.
  • Produced from genetically modified E. coli
  • Monomeric protein, containing 74 amino acids
Eotaxin also negatively regulates adult neurogenesis, and high Eotaxin levels are associated with impaired learning and memory.
Información de suministro: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Precaución: This product is for research use only and is not for use in humans.