Human Recombinant IL-5 (from E. coli)

Proveedor: Shenandoah Biotechnology

100-191-100UG 100-191-10UG 100-191-1MG 100-191-500UG
SHBT100-191-100UGEA 584 EUR
SHBT100-191-100UG SHBT100-191-10UG SHBT100-191-1MG SHBT100-191-500UG
Human Recombinant IL-5 (from E. coli)
Proteínas y Péptidos

Interleukin 5 (IL-5) is a hematopoietic growth factor that is expressed in type 2 T helper (Th2) cells, mast cells, and eosinophils. IL-5 acts through the IL-5 receptor (IL-5R), stimulates B cell growth, and mediates eosinophil activation. Human and mouse IL-5 show cross-reactivity.


  • High Quality
  • Low Endotoxin
  • Affordable


Precaución: For research use only.

Order Now

ESPECIFICACIONES

Nombre de proteína / péptido IL-5
Environmentally Preferable
Sinónimos de proteínas Eosinophil differentiation factor|TRF|B-cell differentiation factor I
Tipo de proteína / péptido Recombinant
Función de la proteína Cytokine
Especies Human
Fuente E. coli
Conjugado Unconjugated
ID del gen P05113
Sequence MIPTEIPTSA LVKETLALLS THRTLLIANE TLRIPVPVHK NHQLCTEEIF QGIGTLESQT VQGGTVERLF KNLSLIKKYI DGQKKKCGEE RRRVNQFLDY LQEFLGVMNT EWIIES
Sin proteasa N
Libre de animales N
Sin nucleasas N
Sin transportista Y
Sin endotoxina N
Nivel de endotoxina Low
Contenido de endotoxinas Endotoxin LAL (EU/µg) ≤0,1
Actividad biológica Bioactivity assay: TF-1 cell proliferation, ED50 ≤0,25 ng/ml, Bioactivity ≥4,0×10⁶ units/mg
Pureza ≥95%
Grado RUO
Forma Lyophilised
Formulación 10 mM sodium glycinate, pH 8,5
Molecular weight 13,3/26,6 kDa
Instrucciones de reconstitución Sterile water at 0.1 mg/ml
Condiciones de almacenamiento –20 °C
Shipping temperature RT
Tested applications Bioassay

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR