Human Recombinant NRG1-B (from E. coli)
Proveedor: Shenandoah Biotechnology
100-46-100UG 100-46-10UG 100-46-1MG 100-46-500UG 100-46-50UG
SHBT100-46-100UGEA 327 EUR
SHBT100-46-100UG SHBT100-46-10UG SHBT100-46-1MG SHBT100-46-500UG SHBT100-46-50UG
Human Recombinant NRG1-B (from E. coli)
Proteínas y Péptidos
Neuregulin 1-beta (NRG1-β) is one of many alternatively-spliced isoforms of the NRG1 gene and contains a soluble EGF-like domain. The EGF-like domain of NRG1-β signals through the ErbB2, ErbB3, and ErbB4 receptor tyrosine kinases. NRG1-β is an important growth factor involved in neuroinflammation, nerve regeneration, and cardiovascular processes.
  • High quality
  • Low endotoxin
  • Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
Información de suministro: Certificate of Analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Precaución: For research use only.