Mouse Recombinant IL-16 (from E. coli)
Proveedor: Shenandoah Biotechnology
200-49-100UG 200-49-10UG 200-49-1MG 200-49-500UG
SHBT200-49-100UGEA 669 EUR
SHBT200-49-100UG SHBT200-49-10UG SHBT200-49-1MG SHBT200-49-500UG
Mouse Recombinant IL-16 (from E. coli)
Proteínas y Péptidos
Interleukin 16 (IL-16) is produced by CD4+ and CD8+ T cells and functions as a chemoattractant for lymphocytes, monocytes, eosinophils, dendritic cells, and Langerhans cells.  Additionally, IL-16 upregulates the expression of the interleukin 2 receptor (CD25), modulates T lymphocyte growth, and suppresses the replication of the human immunodeficiency virus (HIV) and simian immunodeficiency viruses (SIV).
  • High Quality
  • Low Endotoxin
  • Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  
Precaución: For research use only.