Film applicators, automatic, standard
Proveedor: Industrial Physics
AB4120 AB4220 AB4320 AB4400 AB4420
SIMEAB4120EA 7290 EUR
SIMEAB4120 SIMEAB4220 SIMEAB4320 SIMEAB4400 SIMEAB4420
Film applicators, automatic, standard
Aplicadores de película Aplicadores de película de pintura/tinta
The Automatic Film Applicator Standard provides a reliable basis to apply coating films to test charts, panels or foils in a uniform and reproducible way in order to eliminate variations caused by human factors.
  • Paper clamp/vacuum manually operable
  • Full colour display
  • Triple i control (intelligent illumination interface) and easy to update software
  • Drying time recorder option
  • Charts up to A3 size
  • Preset range option
  • Compatible with a large range of application tools
Reproducible
Variations in speed, pressure and direction of draw down causeMore Product Information