Sling psychrometer
Proveedor: Industrial Physics
TM0081
SIMETM0081EA 151.63 EUR
SIMETM0081
Sling psychrometer
Higrómetros Psicrómetros
The psychrometer is designed to measure the percent relative humidity on the basis of the wet bulb-dry bulb thermometer principle.
  • Conversion scale
  • Plastic case on the instrument
  • Relative humidity percent calculations