Tissue processing/embedding cassettes, Unisette™, M405
Proveedor: Simport Scientific
M405-3 M405-4 M405-5 M405-6 M405-7 M405-8 M405-9 M405-10 M405-11 M405-12 M405-3T M405-4T M405-5T M405-6T M405-7T M405-8T M405-9T M405-10T M405-11T M405-12T M405-2T M405-2
SIMPM405-3EA 199 EUR
SIMPM405-3 SIMPM405-4 SIMPM405-5 SIMPM405-6 SIMPM405-7 SIMPM405-8 SIMPM405-9 SIMPM405-10 SIMPM405-11 SIMPM405-12 720-2961 720-2962 720-2963 720-2964 SIMPM405-7T SIMPM405-8T SIMPM405-9T SIMPM405-10T SIMPM405-11T SIMPM405-12T 720-2960 SIMPM405-2
Tissue processing/embedding cassettes, Unisette™, M405
Casetes para histología Casetes para procesamiento de tejidos
These cassettes are moulded from a special high density polymer and keep specimens safely submerged in liquid.
  • Resistant to the chemical action of most histological solvents
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage
  • One-piece integral lid eliminates the need for separate steel lids
  • Snap latch and hinge-lock design prevent early separation of base and lid
Lids can be opened and closed as often as necessary and they always relock securely without danger of specimen loss.
Información para pedidos: Available in non-More Product Information