Placas de pocillos profundos, 384 pocillos cuadrados, Bioblock™
Proveedor: Simport Scientific
T110-200 T110-7
SIMPT110-200EA 279 EUR
SIMPT110-200 SIMPT110-7
Placas de pocillos profundos, 384 pocillos cuadrados, Bioblock™
Microplacas Placas Deep Well
PP. This 384-well plate is ideal for compound storage and handling of biological samples.
  • Round well bottom facilitates aspiration
  • Suitable to be used with DMSO and biological buffers
Información de suministro: Packed in a bag of 4 plates.