Screw cap with septum for sample tubes
Proveedor: Simport Scientific
T504AQX
SIMPT504AQXEA 151 EUR
SIMPT504AQX
Screw cap with septum for sample tubes
Tapas Tapones para tubos de ensayo
This aqua colour screw cap incorporates a pierceable septum made of chemically resistant PTFE on the outside and silicone on the inside, both components being stable over a broad range of temperatures.
  • Acts as a silicone O-ring for better sample protection
  • Especially made to fit and be used with all Simport® sample tubes
The cap is pierceable with pipette tips as well as with syringe needles.