Sigmacote® siliconizing reagent
Proveedor: SIGMA ALDRICH MICROSCOPY
Peligro
M-Clarity™ quality level = MQ200
SL2-100ML SL2-25ML
SL2-100MLEA 225 EUR
SL2-100ML SL2-25ML
Sigmacote® siliconizing reagent
Sigmacote®
Siliconizing reagent for glass and other surfaces. Sigmacote® is a solution of a chlorinated organopolysiloxane in heptane that readily forms a covalent, microscopically thin film on glass.
Pto. de inflamación: –4 °C
Temperatura de almacenaje: Nevera
UN: 2924
ADR: 8(3),II