Laboratory Glassware Washers, Concept Line GW4260
Proveedor: SMEG
GW4260-SC-000 GW4260-S0-000 GW4260-00-000
SMEGGW4260-SC-000EA 9820 EUR
SMEGGW4260-SC-000 SMEGGW4260-S0-000 SMEGGW4260-00-000
Laboratory Glassware Washers, Concept Line GW4260
Lavadoras de vidrio
The laboratory glassware washer for washing and disinfecting of glassware and various laboratory material and used for washing up to 95 °C - antibacterical thermal disinfection.
  • USB frontal port for connectivity
  • Internal memory for cycles, parameters and alarms datalogging
  • Ready to Wi-Fi and Bluetooth connection
  • Washing chamber and internal door in AISI 316L stainless steel
  • External panels made of AISI 304 stainless steel
  • Two independent washing levels through telescopic guides
  • Forced hot air drying system with air filter 98% DOP
  • Ready for absolute HEPA