Aloe vera (200:1), en polvo
Proveedor: Spectrum Chemical
A1861-12KGBL A1861-50KGBL A1861-100GM A1861-25GM
SPCMA1861-12KGBLEA 17120 EUR
SPCMA1861-12KGBL SPCMA1861-50KGBL SPCMA1861-100GM SPCMA1861-25GM
Aloe vera (200:1), en polvo
Aloe vera
Aloe Vera, 200:1, Powder - Spectrum's freeze dried aloe vera powder is easy to use. Just add 199 parts distilled or deionized water to 1 part aloe vera powder to reconstitute the oil derived from the Aloe Vera plant. It is known for its moisturising and healing qualities and can be used in a wide range of personal care products. It is also a humectant, hair conditioners, and to relieve dandruff and dry scalp.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Temperatura de almacenaje: Ambiente
Número MDL: MFCD04118389
Núm. CAS: 85507-69-3
Order Now
Resultados test especificaciones
Moisture max. 10 %
Microbial Limits max. 100 Cfu/G