N-Acetyl-L-tyrosine

Proveedor: Spectrum Chemical

A2017-1KG A2017-25KGBL A2017-5KGBL
SPCMA2017-1KGEA 4250 EUR
SPCMA2017-1KG SPCMA2017-25KGBL SPCMA2017-5KGBL
N-Acetyl-L-tyrosine
N-Acetyl-L-tyrosine

N-Acetyl-L-tyrosine, also known as 4-hydroxyphenylalanine, is used by cells to synthesise proteins.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₁H₁₃NO₄
Temperatura de almacenaje: Nevera
Número MDL: MFCD00037190
Núm. CAS: 537-55-3

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR