tert-Butyl-4-methoxyphenol (mixture of isomers) ≥98.5% FCC

Proveedor: Spectrum Chemical

Sinónimos: Butylhydroxyanisole

BH104-125GM BH104-2.5KG BH104-45KGBL BH104-500GM
SPCMBH104-125GMEA 156 EUR
SPCMBH104-125GM SPCMBH104-2.5KG SPCMBH104-45KGBL SPCMBH104-500GM
tert-Butyl-4-methoxyphenol (mixture of isomers) ≥98.5% FCC
terc-Butil-4-metoxifenol (mezcla de isómeros)

BHA, FCC is an antioxidant and preservative used in food, food packaging and animal feed. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₁₁H₁₆O₂
MW: 180.25 g/mol
Pto. Ebullición: 264 °C (1013 hPa)
Pto. de fusión: 48…55 °C
Densidad: 1,05 g/cm³ (20 °C)
Pto. de inflamación: 156 °C
Temperatura de almacenaje: Ambiente
Número MDL: MFCD01779059
Núm. CAS: 25013-16-5
EINECS: 246-563-8
Índice Merck: 12,01582

Order Now

Resultados test especificaciones

Assay 98.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR