Dehydrocholic acid ≥98.5% USP

Proveedor: Spectrum Chemical

Sinónimos: (R)-4-((5S,8R,9S,10S,13R,14S,17R)-10,13-Dimethyl-3,7,12-trioxohexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic acid

D1001-1KGBL D1001-5KGBL
SPCMD1001-1KGBLEA 3100 EUR
SPCMD1001-1KGBL SPCMD1001-5KGBL
Dehydrocholic acid ≥98.5% USP
Dehydrocholic acid

Dehydrocholic Acid, USP.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₂₄H₃₄O₅
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00066410
Núm. CAS: 81-23-2

Order Now

Resultados test especificaciones

Assay (Dried Basis) 98.5 - 101.0%
Melting Range (Max. 3 C Range) 231 - 242 °C
Specific Rotation [A]D +29.0 to +32.5
Microbial Limits To Pass Test
Loss On Drying max. 1.0 %

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR