Hierro (II) sulfato heptahidrato ≥99.5%, granular USP

Proveedor: Spectrum Chemical

FE110-500GM FE110-2.5KG
SPCMFE110-500GMEA 151 EUR
SPCMFE110-500GM SPCMFE110-2.5KG
Hierro (II) sulfato heptahidrato ≥99.5%, granular USP
Hierro (II) sulfato heptahidrato

Ferrous Sulfate, Heptahydrate, Granular, USP.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: FeSO₄·7H₂O
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00149719
Núm. CAS: 7782-63-0

Order Now

Resultados test especificaciones

Assay (As Heptahydrate) 99.5 - 104.5 %
Arsenic max.3 ppm
Lead max. 10 ppm
Mercury max. 3 Ug/G
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR