Tiamina clorhidrato ≥98.0% FCC

Proveedor: Spectrum Chemical

T1053-25KGBL T1053-500GM T1053-100GM T1053-1KG
SPCMT1053-25KGBLEA 7270 EUR
SPCMT1053-25KGBL SPCMT1053-500GM SPCMT1053-100GM SPCMT1053-1KG
Tiamina clorhidrato ≥98.0% FCC
Tiamina clorhidrato

Thiamine Hydrochloride, FCC is the hydrochloride salt of vitamin B1 and used to treat or prevent vitamin B1 deficiency. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₁₂H₁₈Cl₂N₄OS
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00012780
Núm. CAS: 67-03-8

Order Now

Resultados test especificaciones

Assay (C₁₂H₁₇ClN₄Os[-HCl) 98.0 - 102.0 %
Color of Solution To Pass Test
pH of a 1 in 100 Solution 2.7 - 3.4
Lead (Pb) max. 2 mg/kg
Nitrate (NO₃) To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR