Hollow fibre filter modules, ME membrane, MidiKros® TC
Proveedor: SPECTRUM LABORATORIES
T02-M10U-06-N T02-M20U-10-S T02-M20U-10-N T02-M20U-06-S T02-M20U-06-N T02-M10U-06-S T06-M10U-06-N T06-M20U-10-S T06-M20U-10-N T06-M20U-06-S T06-M20U-06-N T04-M20U-06-S T06-M10U-06-S T04-M20U-10-N T04-M20U-06-N T04-M10U-06-S T04-M10U-06-N T04-M20U-10-S
SPECT02-M10U-06-NEA 515 EUR
SPECT02-M10U-06-N SPECT02-M20U-10-S SPECT02-M20U-10-N SPECT02-M20U-06-S SPECT02-M20U-06-N SPECT02-M10U-06-S SPECT06-M10U-06-N SPECT06-M20U-10-S SPECT06-M20U-10-N SPECT06-M20U-06-S SPECT06-M20U-06-N SPECT04-M20U-06-S SPECT06-M10U-06-S SPECT04-M20U-10-N SPECT04-M20U-06-N SPECT04-M10U-06-S SPECT04-M10U-06-N SPECT04-M20U-10-S
Hollow fibre filter modules, ME membrane, MidiKros® TC
Filtros Sistemas de flujo tangencial
Hollow fibre filter modules are particularly useful in tangential flow filtration because processes can easily be scaled-up from small volume applications to full-scale production.
  • Hollow fibre membranes are anisotropic in structure
  • Filter housings are engineered to handle higher pressures and autoclaving
  • Low protein binding increases the product yield of each processing run
  • TC filters have 1/2" TC/FLL connections and come in three effective lengths
Tangential flow filtration is used in both microfiltration and ultra-filtration applications. Ultra-More Product Information