Hollow fibre filter modules, PS membrane, MidiKros® TC
Proveedor: SPECTRUM LABORATORIES
T02-S050-05-S T02-S500-05-S T02-S500-05-P T02-S500-05-N T02-S05U-05-S T02-S05U-05-P T02-S050-05-P T02-S05U-05-N T02-S050-05-N T02-S010-05-S T02-S010-05-P T02-S010-05-N T06-S500-05-S T06-S500-05-P T06-S500-05-N T06-S05U-05-N T04-S05U-05-N T06-S05U-05-S T04-S500-05-S T04-S500-05-P T04-S500-05-N T04-S05U-05-P T04-S050-05-S T04-S050-05-P T04-S050-05-N T04-S010-05-S T04-S010-05-P T04-S010-05-N T04-S05U-05-S T06-S050-05-S T06-S050-05-P T06-S050-05-N T06-S010-05-S T06-S010-05-P T06-S05U-05-P T06-S010-05-N
SPECT02-S050-05-SEA 652 EUR
SPECT02-S050-05-S SPECT02-S500-05-S SPECT02-S500-05-P SPECT02-S500-05-N SPECT02-S05U-05-S SPECT02-S05U-05-P SPECT02-S050-05-P SPECT02-S05U-05-N SPECT02-S050-05-N SPECT02-S010-05-S SPECT02-S010-05-P SPECT02-S010-05-N SPECT06-S500-05-S SPECT06-S500-05-P SPECT06-S500-05-N SPECT06-S05U-05-N SPECT04-S05U-05-N SPECT06-S05U-05-S SPECT04-S500-05-S SPECT04-S500-05-P SPECT04-S500-05-N SPECT04-S05U-05-P SPECT04-S050-05-S SPECT04-S050-05-P SPECT04-S050-05-N SPECT04-S010-05-S SPECT04-S010-05-P SPECT04-S010-05-N SPECT04-S05U-05-S SPECT06-S050-05-S SPECT06-S050-05-P SPECT06-S050-05-N SPECT06-S010-05-S SPECT06-S010-05-P SPECT06-S05U-05-P SPECT06-S010-05-N
Hollow fibre filter modules, PS membrane, MidiKros® TC
Filtros Sistemas de flujo tangencial
Hollow fibre filter modules are particularly useful in tangential flow filtration because processes can easily be scaled-up from small volume applications to full-scale production.
  • Hollow fibre membranes are anisotropic in structure
  • The filter housings are engineered to handle higher pressures and autoclaving
  • Low protein binding increases the product yield of each processing run
  • TC filters have ¹/₂" TC/FLL connections and come in three effective lengths
Tangential flow filtration is used in both microfiltration and ultra-filtration applications. Ultra-More Product Information
Order Now
ESPECIFICACIONES
Environmentally Preferable
Entrada/salida conexiones 1/2" Tri-clamp/female Luer lock