ErythroClear™ Magnet
Proveedor: STEMCELL Technologies
01737
STMC01737EA 4251 EUR
STMC01737
ErythroClear™ Magnet
Imánes
Perform total nucleic acid extraction from a variety of sample types using the ErythroClear™ magnet.
  • Magnet for column-free immunomagnetic separation
  • Process up to 16 standard polypropylene microcentrifuge tubes at a time
Perform total nucleic acid extraction from a variety of sample types, including whole blood and cell suspensions (PBMCs, hPSCs, cultured cells), using the ErythroClear™ magnet in combination with EasySep™ reagents. Use the ErythroClear™ magnet to process up to 16 standard polypropylene microcentrifuge tubes at a time (8 samples perMore Product Information