MethoCult™ H4534 Classic Medium without EPO
Proveedor: STEMCELL Technologies
04534
STMC04534EA 727 EUR
STMC04534
MethoCult™ H4534 Classic Medium without EPO
Medios de cultivo celular
Methylcellulose-based medium with recombinant cytokines (without EPO) for human cells. 1×100 ml bottle.
MethoCult™ H4534 Classic Without EPO (MethoCult™ GF H4534) is a complete methylcellulose-based medium for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of human bone marrow, mobilised peripheral blood, peripheral blood, and cord blood samples. MethoCult™ H4534 Classic Without EPO is formulated to support the optimal growth of granulocyte-macrophage progenitor cells (CFU-GM, CFU-G and CFU-M).
Precaución: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').