NeuroCult™ Chemical Dissociation Kit (Mouse)
Proveedor: STEMCELL Technologies
05707
STMC05707EA 231 EUR
STMC05707
NeuroCult™ Chemical Dissociation Kit (Mouse)
Suplementos y aditivos de cultivo celular
Kit for chemical dissociation of mouse neurospheres.
  • Obtain cleaner cultures with non-mechanical and non-enzymatic cell dissociation
  • Achieve significantly higher cell viability and total cell number after expansion compared to trituration
  • Ensure minimal cell damage and maintained functional properties of cells
Dissociate neurospheres derived from embryonic or adult mouse central nervous system tissue with NeuroCult™ Chemical dissociation kit. This gentle, non-mechanical and non-enzymatic dissociation kit results in minimal damage to the cells, and ensures higher cell variability and total cell number after expansion in comparison with trituration. Functional properties of cells dissociated with the NeuroCult™ Chemical dissociation kit are also maintained upon subsequent subculture.
Precaución: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').